Supelco® Cerilliant® Organic Reference Standard, 5α-Dihydrotestosterone (DHT) solution
D-073-1ML
: Merck
:
- Concentration:1,0 mg/ml in methanol
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- CAS No.:521-18-6
- Description:5α-Dihydrotestosterone (DHT) solution
- Environmentally Preferable:
- Storage conditions:–70 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:WICE
- MDL No.:MFCD00003667
- M-Clarity quality level:MQ300
- Synonym(s):5α-Dihydrotestosterone solution; 5-alpha-Dihydrotestosterone; DHT; Dihydrotestosterone; 5α−DHT solution
- Molecular weight:290,44
- Flash point:9,7 °C - Closed cup
- Pk Info:Ampule of 1 ml
- GHS signal: GHS02, GHS06, GHS08
: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311