Order Entry
Northern Ireland
ContactUsLinkComponent
 
Diethyl ether ≥99.5%, AR®, Macron Fine Chemicals™
  6783-25
 :  
undefined
Diethyl ether ≥99.5%, AR®, Macron Fine Chemicals™
  6783-25
 :  

 

  • Pk:
    2,5 L
  • Seq:
    5096
  • Environmentally Preferable:

 

 

  • Assay (GC):
    min 99.5%
  • Ethanol:
    max. 0.1%
  • Residue after Evaporation:
    max. 0.03%
  • Stabiliser (BHT):
    ∼5 ppm
  • Titrable Acid (meq/g):
    max. 0.0005
  • Water (KF):
    max. 0.2%

 

 

 : Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
 :
 :74.12 g/mol
 :60-29-7
 :(CH₃CH₂)₂O
 :MFCD00011646
 :35
 :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
 :-123
 :–40 °C
 :12,03852