Genesys Visible and UV/Visible spectrophotometers provide both exceptional performance with a contemporary design.
- Start immediately with the simple to use touch screen start menu
- Exchange samples with ease via large and accessible sample compartment
- Save and print your data fast any way you want it with multiple data export options
- Genesys 140, 150, 180 and BioMate 160 are Wi-Fi compatible with optional USB adapter
The Biomate 160 includes Life Science application software package. Software includes pre-programmed life science methods such as Nucleic Acid A260; Protein A280 and A205; Protein Colorimetric assays and OD600.
The Genesys 180 incudes double-beam optics, and is an ideal solution when a reference beam is required, such as for kinetics. It includes an automated eight-cell changer as standard.
Genesys spectrophotometers provide accurate, reproducible results for your application requirements. Many important applications or experiments can be performed using the visible region of the spectrum including simple enzymatic assays, colour determination and concentration measurements.
These scanning spectrophotometers offer robust, reliable operation in an easy to use and compact design. Whether application is quantitative analysis, QA/QC, life science or education, water and wastewater or food and beverage, you can count on the Genesys instruments.
- Catalog No:
- 634-1075
- 634-1073
- 634-1079
- 634-1083
- SPET840-300000
- Model:
- Genesys™ 50
- Genesys™ 40
- Genesys™ 150
- Biomate™ 160
- Genesys™ 150
- Display:
- 7" high definition colour touch screen
- 7" high definition colour touch screen
- 7" high definition colour touch screen
- 7" high definition colour touch screen
- 7" high definition colour touch screen
- Optical design:
- Dual beam
- Dual beam
- Dual beam
- Dual beam
- Dual beam
- Absorbance range:
- –2 - 3,5 A
- –3 - 3,5 A
- –2 - 3,5 A
- –2 - 3,5 A
- –2 - 3,5 A
- Absorbance accuracy:
- ±0,001 A at 1 A at 546 nm
- ±0,001 A at 1 A at 546 nm
- ±0,001 A at 1 A at 546 nm
- ±0,001 A at 1 A at 546 nm
- ±0,001 A at 1 A at 546 nm
- Photometric accuracy:
- ±0.002 A at 0.5 A;
±0.004 A at 1.0 A;
±0.008 A at 2.0 A
- Wavelength accuracy:
- ±0,5 nm
- ±0,5 nm
- ±0,5 nm
- ±0,5 nm
- ±0,5 nm
- Wavelength repeatability:
- <±0,2 nm
- <±0,2 nm
- <±0,2 nm
- <±0,2 nm
- <±0,2 nm
- Detector:
- Dual-silicon photo diodes
- Dual-silicon photo diodes
- Dual-silicon photo diodes
- Dual-silicon photo diodes
- Dual-silicon photo diodes
- Electrical:
- 100 - 240 V / 50 - 60 Hz
- 100 - 240 V / 50 - 60 Hz
- 100 - 240 V / 50 - 60 Hz
- 100 - 240 V / 50 - 60 Hz
- 100 - 240 V / 50 - 60 Hz
- Lamp source, lifetime:
- Over 5 years typically, 3 years guaranteed
- Over 1000 hours
- Over 5 years typically, 3 years guaranteed
- Over 5 years typically, 3 years guaranteed
- Over 5 years typically, 3 years guaranteed
- Light source:
- Xenon flash
- Tungsten halogen
- Xenon flash
- Xenon flash
- Xenon flash
- Repeatability:
- Photometric: ±0.001 A at 1 A (Measured at 1.0 A at 546 nm)
- Spectral bandwidth:
- 2 nm
- 5 nm
- 2 nm
- 2 nm
- 2 nm
- Stray light:
- <1,0% T 198 nm (KCl) , <0,05% T at 220 nm (NaI), <0,03% T at 340 nm (NaNO2)
- <0,05% T at 340 nm and 400 nm
- <1,0% T 198 nm (KCl) , <0,05% T at 220 nm (NaI), <0,03% T at 340 nm (NaNO2)
- <1,0% T 198 nm (KCl) , <0,05% T at 220 nm (NaI), <0,03% T at 340 nm (NaNO2)
- <1,0% T 198 nm (KCl) , <0,05% T at 220 nm (NaI), <0,03% T at 340 nm (NaNO2)
- W×D×H:
- 355×385×195 mm
- 355×385×195 mm
- 355×385×195 mm
- 355×385×195 mm
- 355×385×195 mm
- Weight:
- 7,5 kg
- 7,5 kg
- 7,5 kg
- 7,5 kg
- 7,5 kg