Aluminium oxide, aerosol refractory paint
Catalog # 40391.9A
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pk:12 oz
- Seq:8480
- Environmentally Preferable:
Specifications
About this item
Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide
:
:101.96 g/mol
Melting Pt:2045
EINECS:215-691-6
UN:1950
REACH:01-2119529248-35
:GHS02
:GHS07
Storage Temperature:Ambient
CAS Number:1344-28-1
:11092-32-3
Formula:Al₂O₃
MDL Number:MFCD00003424
Boiling Pt:–44 °C
Density:3.94 g/cm³ (20 °C)
Flash Pt:–104 °C (–155 °F)
ADR:2.1
Merck Index:13,00355