Potassium dihydrogen phosphate, EMSURE® ISO for analysis, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Monopotassiumphosphate, prim.-Potassium phosphate, Monopotassium phosphate, Potassium phosphate monobasic, Monobasic Potassium Phosphate, Potassium dihydrogen orthophosphate
CAS Number:7778-77-0
:
Formula:KH₂PO₄
MDL Number:MFCD00011401
Boiling Pt:∼100 °C (1013 hPa)
Melting Pt:253
Density:2.3 g/cm³ (20 °C)
Molecular Weight:136.09 g/mol
Merck Index:13,07744
:M-Clarity™ quality level = MQ300
Storage Temperature:Ambient
EINECS:231-913-4
REACH:01-2119490224-41
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Solubility:222 g/l (20 °C)
- Melting Point:253 °C (decomposition)
- Molar Mass:136.08 g/mol
- Bulk Density:1200 kg/m3
- Density:2.34 g/cm3 (20 °C)
- pH:4.4 (50 g/l, H2O, 20 °C)
Specification Test Results
Related Information
Certificates
Search by Lot NumberMERC1.04873.0250_TDS_EN MERC1.04873.1000_TDS_EN ProductPage_104873
Product specification
Merck Individual products and tailor-made chemicals for your technical applications
Product Literature
msds_104873
SDS
Frequently Bought Together