Order Entry
Northern Ireland
ContactUsLinkComponent
 
tert-Butyl methyl ether ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
 :  Merck
 
undefined
tert-Butyl methyl ether ≥99.8%, SupraSolv® for GC-ECD/FID, Supelco®
 :  Merck
 
 
 

 

  • Environmentally Preferable:

 

 

  • Purity (GC):
    =99.8%
  • Identity(IR):
    Conforms
  • Evaporation Residue:
    = 3.0 mg/l
  • Water:
    = 0.02%
  • Color:
    =10 Hazen
  • GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)):
    = 3pg/ml
  • GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)):
    = 3ng/ml
  • Suitability:
    Residue Analysis

 

Frequently Bought Together