Methyl pyruvate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Pyruvic acid ethyl mester, 2-Ketopropionic acid methyl ester, 2-Oxopropionic acid methyl ester, Methyl 2-oxopropanoate, α-Ketopropionic acid methyl ester
CAS Number:600-22-6
:
Formula:C₄H₆O₃
MDL Number:MFCD00008754
Boiling Pt:135…137 °C
Density:1.123
Molecular Weight:102.09 g/mol
Flash Pt:39 °C (102 °F)
Storage Temperature:Refrigerator
EINECS:209-987-4
UN:3272
ADR:3 III
:GHS02
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together