Propylene oxide 99%, pure
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: (±)-Methyloxirane, (±)-Propene oxide, (±)-Propylene oxide
CAS Number:75-56-9
:
Formula:C₃H₆O
MDL Number:MFCD00005126
Boiling Pt:34.3
Melting Pt:–112 °C
Density:0.83 g/cm³
Molecular Weight:58.08 g/mol
Flash Pt:–37 °C
Storage Temperature:Refrigerator
UN:1280
ADR:3 I
Merck Index:12,08041
EINECS:200-879-2
:GHS02
:GHS06
:GHS08
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3650 to 1.3670 (20°C, 589 nm)
- Water:≤0.02 %
- GC:≥98.5 %
- Color scale:≤10 APHA
- Aldehyde (C3H6O):≤0.01 %
Specification Test Results
Frequently Bought Together