Nickel acetate tetrahydrate 99+% for analysis
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Nickel diacetate tetrahydrate, Nickel (II) acetate tetrahydrate, Nickel di(acetate) tetrahydrate, Acetic acid nickel (II) salt tetrahydrate
CAS Number:6018-89-9
:
Formula:C₄H₆NiO₄·4H₂O
MDL Number:MFCD00066973
Boiling Pt:16
Molecular Weight:248.84 g/mol
Melting Pt:250 °C
ADR:9 III
Merck Index:12,06583
EINECS:206-761-7
UN:3077
:GHS07
:GHS08
:GHS09
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Green to blue Crystalline powder
- Infrared spectrum:Conforms
- Titration Complexometric:≥99.0 %
- Aluminium (Al):≤5 ppm
- Cadmium (Cd):≤50 ppm
- Calcium (Ca):≤100 ppm
- Chromium (Cr):≤50 ppm
- Cobalt (Co):≤2000 ppm
- Copper (Cu):≤50 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤50 ppm
- Zinc (Zn):≤50 ppm
Specification Test Results
Related Information
Frequently Bought Together