Order Entry
Northern Ireland
ContactUsLinkComponent
Copper(II) acetate monohydrate, EMSURE® for analysis, Supelco®
Copper(II) acetate monohydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck
Certificates
Copper(II) acetate monohydrate, EMSURE® for analysis, Supelco®
Supplier:  Merck

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Solubility:
    72 g/l (20 °C)
  • Melting Point:
    115 °C
  • Molar Mass:
    199.65 g/mol
  • Bulk Density:
    1100 kg/m3

Specification Test Results

Related Information

Frequently Bought Together