Iodine 0.05 mol/l (0.1 N) in aqueous solution, AVS TITRINORM volumetric solution
Supplier: VWR Chemicals
Certificates
About this item
CAS Number:7553-56-2
:
Formula:I₂
MDL Number:MFCD00011355
Boiling Pt:184.3
Melting Pt:114
Density:1.032 g/cm³ (20 °C)
Molecular Weight:253.81 g/mol
EINECS:231-442-4
ADR:8 II
Merck Index:13,05036
UN:3264
:GHS08
Storage Temperature:Ambient
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.0499 - 0.0501 mol/l
- NIST traceable:Confirmed
Specification Test Results
Related Information
Certificates
Search by Lot Number1L Glass bottle for liquids DIN 45 (ZPACK13415101) 2,5L Glass bottle for liquids DIN 45 (ZPACK13415249)
Packaging Information
spec_VWRC30917.295 spec_VWRC30917.320
Product specification
VWR® Volumetric Solutions for Titration
Product Literature
Frequently Bought Together