Titriplex® III solution c (Na₂-EDTA , 2 H₂O) = 0,1 mol/l, Titripur® Reag. Ph. Eur. for metal titration, Supelco®
Supplier: Merck
Certificates
About this item
Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt
CAS Number:139-33-3
:
Formula:C₁₀H₁₄N₂Na₂O₈
MDL Number:MFCD00070672
Density:1.02 g/cm3 (20 °C)
Molecular Weight:336.21 g/mol
Merck Index:12,03556
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:205-358-3
Limited Time Sale
Select items below are currently on sale
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Form:liquid
- Amount-of-substance concentration:0.0995 - 0.1005
- Measurement uncertainty:+/- 0.0003
- Traceability:NIST SRM
Specification Test Results
Related Information
Certificates
Search by Lot NumberMerck Brighter titration Titripur® Merck Segment Brochure Merck Supelco® Smart Chemical Products Merck Titration Guide for Titer Determination
Product Literature
msds_108431
SDS
ProductPage_108431 spec_Titriplex-III solution Stability data sheet Titripac_108431
Product specification
Frequently Bought Together