Lysis buffer, CTAB
Supplier: PanReac AppliChem
Certificates
About this item
Cationic detergent cetyltrimethylammonium bromide (CTAB) is used to liberate and complex with total cellular nucleic acids.
- Form: Liquid
- Storage temperature: Ambient
- Specification pH (20 °C): 8,0 ±0,1
This buffer contains 2% w/v CTAB, 20 mM EDTA.Na₂.2H₂O, 1,4 M sodium chloride and 100 mM tris ultrapure. Effective for removing polysaccharides, phenolic compounds and other enzyme-inhibiting contaminants found in plant cells in the supernatant.
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together