Restriction enzymes, HindIII, 10 U/μl, Fermentas
About this item
HindIII restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions.
- Convenient color-coded Five Buffer System
- It includes universal Tango buffer for double-digestions
- Reaction buffers contain premixed BSA
- Wide selection of restriction endonuclease specificities
Recognises A^AGCTT sites and cuts best at 37 °C.
3 Options Available Below
Product Details & Documents
HindIII restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions.
- Convenient color-coded Five Buffer System
- It includes universal Tango buffer for double-digestions
- Reaction buffers contain premixed BSA
- Wide selection of restriction endonuclease specificities
Recognises A^AGCTT sites and cuts best at 37 °C.
Specifications for Product Family
Specifications
Specifications
- Catalog No:
- ER0501
- ER0502
- ER0505
- Enzyme name:
- Hind III
- Hind III
- Hind III
- Overhang type:
- 5' overhang
- 5' overhang
- 5' overhang
- Compatible buffer:
- FastDigest Green Buffer or FastDigest Buffer (colorless)
- FastDigest Green Buffer or FastDigest Buffer (colorless)
- FastDigest Green Buffer or FastDigest Buffer (colorless)
Specifications
Product Family Options
Product Information
- ConcentrationUnitsPkAvailabilityPrice
- ER0501HindIII, 10 U/μlSpecifications:
More
Specifications
Cat. No.ER0501Enzyme nameHind IIIOverhang type5' overhangCompatible bufferFastDigest Green Buffer or FastDigest Buffer (colorless) - ER0502HindIII, 10 U/μlSpecifications:
More
Specifications
Cat. No.ER0502Enzyme nameHind IIIOverhang type5' overhangCompatible bufferFastDigest Green Buffer or FastDigest Buffer (colorless) - ER0505HindIII, 10 U/μlSpecifications:
More
Specifications
Cat. No.ER0505Enzyme nameHind IIIOverhang type5' overhangCompatible bufferFastDigest Green Buffer or FastDigest Buffer (colorless)