Resveratrol 99%
ACRO430075000
:
- Pack type:Glass bottle
- Pk:500 mg
- Seq:2725
- Environmentally Preferable:
- Appearance:Off-white to beige Powder
- Infrared spectrum:Conforms
- Melting point:257 to 264 °C
- Loss on drying:≤1 %
- HPLC:≥98.5 %
: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol
:501-36-0
:
:C₁₄H₁₂O₃
:MFCD00133799
:228.25 g/mol
:GHS05
:GHS07
:261…263 °C
:Refrigerator
:12,08326