Order Entry
Northern Ireland
ContactUsLinkComponent
3-Methylsalicylaldehyde 98%
3-Methylsalicylaldehyde 98%
Certificates
undefined
3-Methylsalicylaldehyde 98%

Specifications

  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to yellow to orange Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5620 to 1.5650 (20°C, 589 nm)
  • GC:
    ≥97.5 %

Specification Test Results

Related Information

Frequently Bought Together