ContactUsLinkComponent
Polypropylene glycol, M.W. ∼ 425
Polypropylene glycol, M.W. ∼ 425
Certificates
undefined
Polypropylene glycol, M.W. ∼ 425
Specifications
  • Environmentally Preferable:
Specifications
Specification Test Results
  • Appearance:
    Clear viscous liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.15 %
  • Color scale:
    ≤70 APHA
  • Acid value:
    ≤1 mg KOH/g
  • Hydroxyl value:
    250 to 276 mg KOH/g