Order Entry
Northern Ireland
ContactUsLinkComponent
Anti-USE1 Rabbit Polyclonal Antibody
Anti-USE1 Rabbit Polyclonal Antibody
Catalog # ABGEAP19206A
Supplier:  Abgent
Anti-USE1 Rabbit Polyclonal Antibody
Catalog # ABGEAP19206A
Supplier:  Abgent
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    unconventional SNARE in the ER 1 homolog (S. cerevisiae)
  • Antigen symbol:
    USE1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Gene ID:
    55850
  • Antigen synonyms:
    SLT1|D12|MDS032|P31
  • Amino acid number:
    9 to 35
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    29.371 kDa
  • Sequence:
    KRDPDEWRLEKYC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0.37
  • Shipping temperature:
    4 °C
  • Immunogen:
    This USE1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 9-35 amino acids from the N-terminal region of human USE1.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: USE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Mouse