VWR®, Autoclavable Biohazard Bags
Ideal for processing biohazardous materials that require autoclaving prior to disposal. Available with or without temperature indicator. High integrity polypropylene bags withstand 135 °C (275 °F) maximum autoclave temperature. PP, 50 µm thick, yellow.
- Twist-tie closure allows bag to expand and vent steam during cooling
- Bags resist punctures, tears and leaks, and they also meet the 165 g dart-drop standard as required by ASTM D1709-98
- Each bag is imprinted with the universal biohazard symbol
All biohazard bags have a temperature indicator patch with the words 'Contents Have Been Autoclaved' printed in a chemically active ink. Before autoclaving the ink is clear on a white background, after autoclaving is complete, it darkens and words appear in a dark brown colour, providing proof that bag has been exposed to high temperatures for sterilisation.
PP, 50 µm thick, yellow
Frequently Bought Together