Bostwick consistometers
Supplier: ENDECOTTS
Certificates
About this item
Bostwick consistometers are durable stainless steel instrument for accurately checking laboratory or production samples against consistency, viscosity or flow rate standards.
- Consistent platform for test
- Requires up to 100 ml of sample
- Leveling screws and spirit level enable accurate set up
Used in chemical, paint, cosmetic and food processing industries.
Certifications: R-81294D and ASTM F1080-93
Frequently Bought Together