Reverse osmosis systems, arium® advance RO
Directly connected to your potable feed water source, this system delivers pure water (Type III) of the highest quality.
- Touch-function display and intuitive menu navigation for error-free control, even while wearing gloves
- iJust software: Enhances the quality of purified water, reduces water usage and calculates the optimal cleaning cycles for the system
- Produced water is stored in the unique, closed arium® bagtank system (not included), which reliably protects purified water from secondary contamination
- Innovative Bag technology eliminates costly and time-consuming tank cleaning
- Can be used as a bench-top, wall-mounted or a built-in unit
Typical applications include: Purification of feed water used to supply ultrapure water systems, rinsing glassware and laboratory vessels, purification of feed water for various laboratory instruments, such as humidifiers, autoclaves and glassware dishwashers.
: Bagtanks and bags have to be ordered separately.
- Catalog No:
- VIVAH2O-RO-2-T
- VIVAH2O-RO-2-B
- Model:
- arium® advance RO
- arium® advance RO
- Feed water:
- Potable mains water supply
- Potable mains water supply
- Production flow rate:
- 16 L/h
- 16 L/h
- Particles:
- >99% rejection
- >99% rejection
- W×D×H:
- 350×451×501 mm
- 350×451×501 mm
- Weight:
- 15 kg
- 15 kg
Frequently Bought Together