Order Entry
Northern Ireland
ContactUsLinkComponent
Sartobind® Filtration Capsules
Sartobind® Filtration Capsules
Supplier:  Sartorius
Certificates
undefined
Sartobind® Filtration Capsules
Supplier:  Sartorius
Certificates

Specifications

  • Catalog No:
  • SARP92STPA42DD-11D
  • SARP92IEXQ42D4-OOA
  • SARP92IEXQ42DD-11D
  • SARP96HICP42EUC11A
  • SARP92IEXS42D4-SSA
  • SARP92IEXS42D4-OOA
  • SARP92IEXQ42D4-SSA
  • SARP92HICP42DD-11D
  • Type:
  • Sartobind STIC® PA Pico
  • Sartobind® Q SingleSep Mini
  • Sartobind® Q Pico
  • Sartobind® Phenyl Nano
  • Sartobind® S SingleSep Mini
  • Sartobind® S SingleSep Mini
  • Sartobind® Q SingleSep Mini
  • Sartobind® Phenyl Pico
  • Water flow rate:
  • 0,8 - 2,4 ml/min
  • 140 ml/min
  • 0,8 - 2,4 ml/min
  • 15 ml/min
  • 140 ml/min
  • 140 ml/min
  • 140 ml/min
  • 0,8 - 2,4 ml/min
  • Filtration area:
  • 2,9 cm²
  • 250 cm²
  • 2,9 cm²
  • 110 cm²
  • 250 cm²
  • 250 cm²
  • 250 cm²
  • 2,9 cm²
  • Inlet/outlet:
  • Luer female connectors
  • Hose barb connectors
  • Luer female connectors
  • Luer female connectors
  • Sanitary 1/2" - 3/4"
  • Hose barb connectors
  • Sanitary 1/2" - 3/4"
  • Luer female connectors
  • Ø×L:
  • 11×31 mm
  • 45×103 mm
  • 11×31 mm
  • 33×37 mm
  • 45×87 mm
  • 45×103 mm
  • 45×87 mm
  • 11×31 mm
  • Max. pressure:
  • 6 bar
  • 4 bar
  • 6 bar
  • 4 bar
  • 4 bar
  • 4 bar
  • 4 bar
  • 6 bar

Specifications

Related Information

Frequently Bought Together