SP Bel-Art Cleanware™ Bottle Brush, Bel-Art Products, a part of SP
13196-072
- Bristle Material:Hog hair
- Handle Material:Wood
- Descripción:Bottle brush for necks larger than 1"
- Environmentally Preferable:
- Brush Head Width:21 mm
- Referencia:13196-072
- Longitud del mango:370 mm
- Nº de proveedor:170680000
- Longitud del cepillo:45 mm
- Trim Length:8 mm
Uniquely designed, swiveling pivot head enables thorough cleaning of bottles and flasks including difficult curves and corners.
- Natural bristles set in wood will not scratch glass
- Brush head is slightly convex
- Brush head is attached to hardwood handle
The swivel head fits through a 29/42 standard taper neck, or any non-tapered neck with a diameter of 24 mm (1") or larger.