Order Entry
Mexico
ContactUsLinkComponent
 
Pentair Fibrillated polypropylene string-wound cartridge, 25 µm, 10" length
  MFLX29802-42
 :  Avantor
undefined
Pentair Fibrillated polypropylene string-wound cartridge, 25 µm, 10" length
  MFLX29802-42
 :  Avantor

 

String-wound pattern provides greater surface area for lower pressure drop and higher flow.

  • Contructed of polypropylene (PP)

Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165 °F (74 °C).

Fibrillated cartridges are non-leaching and offer minimal fiber migration with a higher pressure drop.

  • Filter Media:
    PP
  • Micron Rating:
    25
  • Description:
    Fibrillated polypropylene string-wound cartridge, 25 µm, 10" length
  • Environmentally Preferable:
  • Cat. no.:
    MFLX29802-42
  • Filter Length:
    10"
  • Warranty:
    1 year