Order Entry
Mexico
ContactUsLinkComponent
 
Masterflex® Fibrous polypropylene string-wound cartridge, 1 µm, 10" length
  MFLX01508-77
 :  Avantor
undefined
Masterflex® Fibrous polypropylene string-wound cartridge, 1 µm, 10" length
  MFLX01508-77
 :  Avantor

 

String-wound pattern provides greater surface area for lower pressure drop and higher flow.

  • Contructed of polypropylene (PP)

Polypropylene string is resistant to chemical and bacterial attack. Suitable to 165 °F (74 °C).

  • Filter Media:
    PP
  • Micron Rating:
    1
  • Description:
    Fibrous polypropylene string-wound cartridge, 1 µm, 10" length
  • Environmentally Preferable:
  • Cat. no.:
    MFLX01508-77
  • Filter Length:
    10"
  • Warranty:
    1 year