Streptococcus pneumoniae Slide
470177-408
: Avantor
- Description:Streptococcus pneumoniae Slide
- Environmentally Preferable:
- Cat. no.:470177-408
- Stain:Gram (+)
- Supplier No.:WARD470177-408
- Orientation:Smear
Human pathogenic bacteria
- Identifying features clearly distinguishable
- Rigorous quality control standards
Oval to spherical pairs. Agent of pneumonia and meningitis in humans.