Order Entry
Mexico
ContactUsLinkComponent
Grafco® Sprague Rappaport Stethoscope
Grafco® Sprague Rappaport Stethoscope
Catalog # 470003-428
Supplier:  GRAHAM-FIELD INC
undefined
Grafco® Sprague Rappaport Stethoscope
Catalog # 470003-428
Supplier:  GRAHAM-FIELD INC
Supplier Number:  602
Specifications
  • Description:
    Sprague Rappaport Stethoscope
  • Environmentally Preferable:
  • Cat. no.:
    470003-428
  • Supplier No.:
    602
Specifications
About this item
This professional, high-quality Sprague Rappaport type stethoscope has the best sound quality of all available stethoscopes.
  • Five interchangeable chest pieces (two diaphragms, three bells)
  • Ideal for infant, pediatric, and adult use
  • Heavy-walled 22" tubing decreases extraneous sounds
  • Latex-free
The exclusive, advanced design boasts an improved valve mechanism that virtually eliminates acoustic leakage, making it an excellent instrument to demonstrate heartbeat.