Order Entry
Mexico
ContactUsLinkComponent
Cuvette Rack
Cuvette Rack
  470149-208
undefined
Cuvette Rack
  470149-208
  • Description:
    Cuvette Rack
  • Environmentally Preferable:
  • Cat. no.:
    470149-208
  • Supplier No.:
    260010
Accommodate Most Square Cuvettes.
  • Reduced risk of spilling
  • Avoids risk of confusion with individually numbered spots
  • Firmly holds up to 20 individual cuvettes