Order Entry
Mexico
ContactUsLinkComponent
Orion™ Star™ A325 pH/Conductivity Portable Multiparameter Meter, Thermo Scientific
Orion™ Star™ A325 pH/Conductivity Portable Multiparameter Meter, Thermo Scientific
Catalog # 89206-366
undefined
Orion™ Star™ A325 pH/Conductivity Portable Multiparameter Meter, Thermo Scientific
Catalog # 89206-366
Supplier Number:  STARA3255
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Calibration:
    pH with calibration editing option, relative mV (RmV), ORP, conductivity with calibration editing option and temperature
  • Memory:
    Non-volatile memory preserves data log, calibration log, and meter settings
  • No. of channels:
    2
  • Temperature range:
    –5 to 105°C (22 to 221°F)
  • Description:
    Orion™ Star™ A325 pH/conductivity portable meter kit, with ROSS Ultra Triode low maintenance gel pH/ATC electrode and DuraProbe 4-cell conductivity sensor
  • Environmentally Preferable:
  • Operating temperature range:
    5…45 °C (41…113 °F)
  • Temperature accuracy:
    ±0.1°C
  • Temperature resolution:
    0.1°C, 0.1°F
  • Resistivity resolution:
    1 ohm or 0.1 meg-ohm, auto ranging
  • Battery life:
    800 h
  • Input:
    BNC (pH or ORP electrode), pin-tip (reference electrode), 8 pin MiniDIN (conductivity sensor with built-in temperature or ATC temperature probe)
  • IP rating:
    IP-67
  • Alarm:
    High/low limit alarm, calibration due alarm
  • Calibration points:
    1 point relative mV (RmV) mode; 1 point ORP mode; up to 5 points Conductivity mode; 1 to 5 points pH mode
  • Conductivity accuracy:
    0.5% of reading ±1 digit greater than 3 μS; 0.5% of reading ±0.01 μS less/equal than 3 μS
  • Conductivity cell constant:
    0.001 - 199.9 cm⁻¹
  • Conductivity range:
    0.001 µS/cm - 3000 mS/cm
  • Conductivity resolution:
    0.001 μS minimum, auto ranging up to 4 significant digits
  • Data logging capacity:
    5000 data points with time/date stamp
  • Dimensions:
    24L x 10W x 6.5H cm (9.5 x 3.9 x 2.6")
  • Display:
    Graphic LCD with backlight
  • Languages:
    English, Spanish, French, Italian, German, and Chinese (Portuguese and Korean available with software update)
  • Outputs:
    USB, RS232
  • Power supply:
    4 AA batteries (included) or universal AC adapter (sold separately)
  • Resistivity range:
    2 ohm to 100.0 meg-ohm
  • Resistivity accuracy:
    0.5% of reading ±1 digit
  • Salinity accuracy:
    0.5% of reading ±1 digit
  • Salinity range:
    0.06 to 80.00 psu, 0.05 to 42.00 ppt
  • Salinity resolution:
    0.01 psu or 0.01 ppt minimum, auto ranging
  • TDS accuracy:
    0.5% of reading ±1 digit
  • TDS range:
    0.001 to 200.0 ppm
  • TDS resolution:
    0.001 ppm minimum, auto ranging up to 4 significant digits
  • Weight:
    0.5 kg (1.2 lbs.)
  • mV accuracy:
    ±0.2 mV or ±0.05% of reading whichever is greater
  • mV range:
    ±2000.0 mV
  • mV resolution:
    0.1 mV
  • pH accuracy:
    ±0.002 pH
  • pH range:
    -2.000 to 20.000 pH
  • pH resolution:
    0.1, 0.01, 0.001 pH
  • Battery type:
    Four AA (included)
  • Cat. no.:
    89206-366
  • Includes:
    Star™ A325 pH/conductivity portable meter, Orion ROSS Triode pH/ATC gel-filled electrode with 3m cable, Orion DuraProbe conductivity cell with 3m cable, pH 4.01 buffer pouches, pH 7.00 buffer pouches, pH 10.01 buffer pouches, conductivity standard pouches, rinse solution, protective armor with electrode holder, hard carrying case, and batteries
  • Supplier No.:
    STARA3255

Specifications

About this item

Accurately measure pH, conductivity, and temperature with the Orion™ Star™ A325 pH/Conductivity Portable Meter for high performance in challenging field tests.

  • Combined convenience with the benefits of a pH meter and conductivity meter in one easy-to-use system
  • Measure pH, mV, relative mV, or ORP with temperature on channel one and measure conductivity, TDS, salinity, or resistivity with temperature on channel two
  • Calibration editing option allows individual points to be fixed without a full recalibration
  • Auto-ranging conductivity values offer consistent readings across entire measurement range without need to recalibrate
  • Capture critical readings with measurement stability indicator and selectable read modes
  • Auto-Read mode holds a stable reading, Continuous mode shows changing readings with hold option, and Timed mode logs data at specific intervals
  • Three-year meter warranty

Get high accuracy and premium performance everywhere needed with the Orion™ Star™ A325 pH/Conductivity Portable Meter, designed for a wide range of pH, mV, ORP, conductivity, total dissolved solids (TDS), salinity, resistivity, and temperature testing and field applications.

Take measurements at the source with this premium line of portable meters. The Orion™ Star™ A325 pH/Conductivity Portable Meter offers an information-rich screen and user-friendly operation. This rugged, waterproof portable meter is ready to tackle the elements. These meters are designed to be easy to use, dependable field meters that offer advanced features and functions, including a 5000 point date log with sample ID and user ID options, calibration and high/low limit alarms, and password-protected measurement methods. Easily view individual measurements on a full screen display and quickly toggle to a split screen display to view multiple measurements simultaneously. Perform up to a five point pH calibration using automatic buffer recognition of NIST or DIN buffer groups with choice to manually enter custom buffer values. Perform up to a five point conductivity calibration. Use linear, non-linear, EP (USP), or off conductivity temperature compensation with 5, 10, 15, 20, or 25 °C reference temperatures. Flexible power options with AA batteries or optional universal power adapter. Non-volatile meter memory ensures data and settings are protected, even with loss of power.

Evaluate samples in the most demanding locations using this waterproof meter with IP67-rated housing. Use the two channels to measure pH and conductivity simultaneously or view each channel separately. Log up to 5000 data point sets with time/date stamp and transfer logs to a printer or computer.

Certifications: CE, TUV 3-1, FCC Class A.
Ordering information: For compatible pH electrode, see Orion™ item 8107UWMMD. For compatible conductivity probe, see Orion™ item 013010MD. For compatible buffer solutions and kits, see Orion™ items 910410-WA, 910710, and 911010.