VWR® Certified Liquid-In-Glass Thermometers
Supplier: VWR International
Certificates
About this item
Organic liquid. White back. Accurate to the requirements of ANSI/SAMA Z236.1-1983, ASTM, NIST, DKD and/or equivalent standards. Feature a Calibration Report stating corrections to one-tenth of the smallest division and individual serial number.
- Individually serialized
- Lead-free glass
Ideal for use in chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical/medical industry, manufacturing, dairies, breweries and distilleries.
Certifications: NIST.
Ordering information: Includes individual Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration.
Related Information
Certificates
Search by Lot NumberAT-VWR Thermometer FAQs
Datasheet
VWR GHS Compliance Letter (US)
Additional Product Details
Frequently Bought Together