Order Entry
Mexico
ContactUsLinkComponent
Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Catalog # 10806-000
undefined
Puritan® Swabs, Cotton Head, Wood Shaft, Puritan Medical Products
Catalog # 10806-000
Supplier Number:  806-WC

Specifications

  • Description:
    With adhesive
  • Environmentally Preferable:
  • Cat. no.:
    10806-000
  • Handle Length:
    152.4 mm (6")
  • Head Width:
    4.7 mm (0.188")
  • Head Length:
    15.9 mm (0.625")
  • Handle Width:
    2.5 mm (0.1")
  • Supplier No.:
    806-WC

Specifications

About this item

These Puritan® swabs are ideal for use in bacteriology labs for inoculating broth and plated media, and obtaining culture specimens.

Swabs with adhesive incorporate a nonsterile cotton tip secured by adhesive to the wooden handle. Swabs are also available without adhesive, for use where contamination of critical surfaces is a concern. Swabs are autoclavable.

They are also useful for cleaning delicate laboratory instruments.

Ordering information: Packaged in bags of 100 swabs, 10 bags to a pack, 10 packs to a case.