Diethyl ether, anhydrous (max. 0.001% H₂O) ≥99.5% (by GC, corrected for water content) stabilised for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
: Avantor
: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
:60-29-7
:
:(CH₃CH₂)₂O
:MFCD00011646
:35
:0.7135 g/cm³ (20 °C)
:1.5 - 3.5% Stabiliser
:74.12 g/mol
:-123
:-40
:200-467-2
:1155
:3 I
:12,03852
:Management of Change (MOC) category = NI
:01-2119535785-29
:GHS02
:GHS07
:GHS08
:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Environmentally Preferable:
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400-350 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.07
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 231 nm:≤0.4
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤220
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.5 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Assay (by GC, corrected for water, exclusive of preservative):≥99.5 %
- Peroxide (as H₂O₂):≤5 ppm
- Preservative (C₂H₅OH):1.5 - 3.5 %
- Residue after Evaporation:≤5 ppm
- Titrable Acid (µeq/g):≤0.2
- Water (H₂O)(by coulometry):≤0.01 %
Frequently Bought Together