CleanMax® Clean and Sterile Disposable Hoods, Lakeland Industries
Catalog # 76193-232
Supplier: CHEMLAND-LAKELAND INDUS
Specifications
- Clothing Size:One size
- Sterile:+
- Garment application:Cleanroom
- Gender:
- Color:White
- Description:Clean Sterile Hood
- Environmentally Preferable:
- Cat. no.:76193-232
Specifications
About this item
Clean and Sterile disposable hoods are individually double-bagged and folded to promote easier gowning with less excursions.
- Clean and sterile in silicone-free environment
- Latex-free
- Covers shoulders
- One size
- Ties to customize fit
- Individually double-bagged
- Gamma radiation indicator dots on each package
CleanMax® cleanroom garments provide comfort, quality, and protection.
All these apparels provide chemical penetration resistance to oils, bleach, and 50% sodium hydroxide. CleanMax® apparel is also resistant to blood and body fluid penetration, viral penetration, and blood borne pathogens.
CleanMax® garments feature bound seams, which are precisely sewn with an additional outer binding. This increases seam strength and provides a better barrier from particulates than simple serged seams. The tunneled elastic is completely covered by the garment material so that the elastic is not exposed to the work environment.
Certifications: Compatible for ISO Class 5 – 8 Cleanrooms and all Controlled Environments.