Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)
Catalog # 95017-134
Supplier: Cytiva
Product available on GSA Advantage®
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:500 g
- Environmentally Preferable:
- Cat. no.:95017-134
- Supplier No.:17-1321-01
Specifications
Specification Test Results
- Application:For preparing electrophoresis buffer. Used in PAGE and Sequencing.
- Purity:≥99,8%
- pH Range:7.2 - 9.0
- Boiling/Condensation Point:219 - 220°C
- Melting/Freezing Point:170°C
- Solubility:Soluble in cold water and hot water, methanol
- Physical State:Solid
- Color:White
- Storage conditions:10 - 30°C
- Estimated shelf life from manufacture date:12 month
- Wetted Part:No
- Guidance:Tris = (tris(hydroximethyl)aminoethane)
- DIGE approved:Y
Specification Test Results
About this item
Synonyms: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM
:
:121.14 g/mol
Boiling Pt:357
EINECS:201-064-4
:GHS07
Melting Pt:169…172 °C
Storage Temperature:Ambient
CAS Number:77-86-1
Formula:NH₂C(CH₂OH)₃
MDL Number:MFCD00004679
Density:1.328 g/cm³ (20 °C)
Merck Index:13,09842