Order Entry
Mexico
ContactUsLinkComponent
Eisco® Urinary Bladder with Prostate Model
Eisco® Urinary Bladder with Prostate Model
Catalog # 470136-164
undefined
Eisco® Urinary Bladder with Prostate Model
Catalog # 470136-164
Supplier Number:  AM0113

Specifications

  • Description:
    Urinary bladder with prostate
  • Height:
    16 cm
  • Length:
    21 cm
  • Environmentally Preferable:
  • Cat. no.:
    470136-164
  • Width:
    21 cm
  • Supplier No.:
    AM0113
  • No. of Parts:
    2

Specifications

About this item

Reveal internal structures of bladder and prostate.

  • Model is sectioned medially for detailed examination
  • Designed by medical professionals and educators
  • Custom molded using the highest quality nontoxic polymers and PVC
  • Hand-painted by skilled artists
  • Includes a key card

Enlarged three times, shows the male urinary bladder with the prostate gland surrounding the urethra. The model is dissected medially to expose both internal and external structures of the bladder and prostate, including the ureteric and urethral orifices, ducts deferens, seminal gland, ejaculatory duct.