Order Entry
Mexico
ContactUsLinkComponent
VECTASHIELD® Mounting Medium, Vector Laboratories
  101098-042
 :  
undefined
VECTASHIELD® Mounting Medium, Vector Laboratories
  101098-042
 :  H-1000-10
 :  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    VECTASHIELD® Mounting Medium
  • Size:
    10 mL
  • Environmentally Preferable:
  • Cat. no.:
    101098-042
  • Supplier No.:
    H-1000
VECTASHIELD® Mounting Media minimize photobleaching during fluorescence-based imaging. There are three versions: original VECTASHIELD, a non-curing formulation that allows immediate slide viewing; VECTASHIELD HardSet, which cures for long-term storage without sealing; and VECTASHIELD Vibrance™, a new hardening formulation that offers superior handling and greater stability for long-term storage of slides at room temperature.
VECTASHIELD® Mounting Media are compatible with fluorescent nuclear stains, fluorescent proteins, fluorescent tracers, histochemical stains, and most fluorochromes, with VECTASHIELD Vibrance showing superior performance with far-red dyes.
All three VECTASHIELD formulations are available with and without DAPI, a common fluorescent nuclear counterstain. DAPI excites at about 360 nm and emits at about 460 nm when bound to DNA, producing nuclei with blue fluorescence. VECTASHIELD HardSet is also available with phalloidin (PI), a bicyclic heptapeptide that stains actin filaments. PI excites at about 535 nm and emits at about 615 nm, producing a red fluorescence.
 : Manufactured under ISO9001:2015