Order Entry
Korea, Republic of
ContactUsLinkComponent
Butyl-4-hydroxybenzoate 98.0-102.0% (dry basis), GenAR® NF, Macron Fine Chemicals™
Butyl-4-hydroxybenzoate 98.0-102.0% (dry basis), GenAR® NF, Macron Fine Chemicals™
Catalog # MACR7558-18
CAS Number:  
Butyl-4-hydroxybenzoate 98.0-102.0% (dry basis), GenAR® NF, Macron Fine Chemicals™
Catalog # MACR7558-18
CAS Number:  

Specifications

  • Pack type:
    Corrugated case
  • Pk:
    25 kg
  • Seq:
    4283
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Bulk Pharmaceutical Chemical:
    98.0 - 102.0%
  • NF - Assay (C₁₁H₁₄O₃):
    98.0 - 102.0 %
  • NF - Identification A:
    Passes Test
  • NF - Identification B:
    Passes Test
  • NF - Color of Solution:
    Passes Test
  • NF - Acidity:
    Passes Test
  • NF - Residue on Ignition:
    ≤0.1 %
  • NF - Related Substances - p-Hydroxybenzoic Acid:
    ≤0.5 %
  • NF - Related Substances - Unspecified Impurities (each):
    ≤0.5 %
  • NF - Related Substances - Total Impurities:
    ≤1.0 %
  • EP/BP - Assay (C₁₁H₁₄O₃):
    98.0 - 102.0 %
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Acidity:
    Passes Test
  • EP/BP - Related Substances - Impurity A:
    ≤0.5 %
  • EP/BP - Related Substances - Unspecified Impurities (each):
    ≤0.5 %
  • EP/BP - Related Substances - Total Impurities:
    ≤1.0 %
  • EP/BP - Sulfated Ash:
    ≤0.1 %
  • JP - Acidity:
    Passes Test
  • JP - Assay:
    98.0 - 102.0 %
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Heavy Metals:
    ≤20 ppm
  • JP - Identification:
    Passes Test
  • JP - Melting Point:
    68 - 71 °C
  • JP-RelatedSubstances- p-Hydroxybenzoic Acid:
    ≤0.5 %
  • JP-Related Substances-Unspecified Impurities (each):
    ≤0.5 %
  • JP-Related Substances-Total Impurities:
    ≤1.0 %
  • JP - Residue on Ignition:
    ≤0.10 %

Specification Test Results

About this item

Synonyms: Butyl 4-hydroxybenzoate, 4-Hydroxybenzoic acid butyl ester, Butyl parahydroxybenzoate, Butylparaben
 :
 :194.23 g/mol
EINECS:202-318-7
CAS Number:94-26-8
Formula:C₁₁H₁₄O₃
MDL Number:MFCD00016478
Boiling Pt:156…157 °C (3.5 mmHg)
Melting Pt:73 °C
Density:1.23 g/cm³ (20 °C)
Merck Index:13,01583