Viral inactivation solution, Biotech Reagent, J.T.Baker®
: Avantor
:9043-30-5
:0.9 – 1.1 g/ml
:Ambient
- Environmentally Preferable:
- ID (by FTIR):Conforms
- Appearance:Passes Test
- Purity by HPLC:min 95%
- Assay:Passes Test
- Acid value:max 10
- Viscosity (25°C):For information only
- Density:0.9 – 1.1 g/mL
- Limit of free ethylene oxide (Target <1 ppm):For information only
- Total Aerobic Microbial Count:1000 cfu/mL
- Total Yeast and Mold Count:10 cfu/mL
- Endotoxin:For information only
- Protein reactive impurities (100x Diluted):For information only
- Arsenic (As):For information only
- Cadmium (Cd):For information only
- Lead (Pb):For information only
- Mercury (Hg):For information only
- Cobalt (Co):For information only
- Nickel (Ni):For information only
- Vanadium (V):For information only
- Silver (Ag):For information only
- Gold (Au):For information only
- Iridium (Ir):For information only
- Osmium (Os):For information only
- Palladium (Pd):For information only
- Platinum (Pt):For information only
- Rhodium (Rh):For information only
- Ruthenium (Ru):For information only
- Selenium (Se):For information only
- Thallium (Tl):For information only
- Barium (Ba):For information only
- Chromium (Cr):For information only
- Copper (Cu):For information only
- Lithium (Li):For information only
- Molybdenum (Mo):For information only
- Antimony (Sb):For information only
- Tin (Sn):For information only
Enhancing Virus Clearance Process Capability using a Biodegradable Detergent J.T Backer VIS Viral Inactivation Study J.T.Baker Viral Inactivation Solution J.T.Baker Viral Inactivation Solution (Detergent) Clearance by Purification Process J.T.Baker Viral Inactivation Solution Ease of Use
Frequently Bought Together