4-Methylcyclohexanol cis- and trans-mixture ≥98.0% (by GC)
: TCI
:589-91-3
:
:C₇H₁₄O
:MFCD00001449
:171
:114.19 g/mol
:-50
:68°C
:209-664-8
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
Frequently Bought Together