2-Phenyl-2-oxazoline ≥98.0% (by GC, titration analysis)
: TCI
: 2-Phenyl(2-oxazoline), (4,5-Dihydro-2-oxazolyl)benzene, 4,5-Dihydro-2-phenyloxazole
:7127-19-7
:
:C₉H₉NO
:MFCD00075541
:243°C
:147.1739 g/mol
:GHS07
:Refrigerator
- Environmentally Preferable:
- Appearance:White or Colorless to Light yellow powder to lump to clear liquid
- Purity (GC):min. 98.0 %
- Purity (Nonaqueous Titration):min. 98.0 %
Frequently Bought Together