Order Entry
Italy
ContactUsLinkComponent
 
cis-4-Methyl-2-pentene ≥93.0% (by GC)
Catalog # TCIAM0393-5ML
Supplier:  TCI
CAS Number:  
undefined
cis-4-Methyl-2-pentene ≥93.0% (by GC)
Catalog # TCIAM0393-5ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

Specifications

Specification Test Results

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 93.0 %

Specification Test Results

About this item

Synonyms: (Z)-4-Methyl-2-pentene
 :
 :84.16 g/mol
EINECS:211-721-7
UN:2288
 :GHS02
 :GHS08
Storage Temperature:Refrigerator
CAS Number:691-38-3
Formula:C₆H₁₂
MDL Number:MFCD00009293
Boiling Pt:56°C
Melting Pt:-134.8
Flash Pt:–33°C
ADR:3 II