3-Methyl-2-hexanone ≥98.0% (by GC)
Catalog # TCIAM0234-10ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 mL
- Seq:2275
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Light yellow clear liquid
- Purity (GC):min. 98.0 %
Specification Test Results
About this item
Synonyms: Methyl 1-methylbutyl ketone, 3-Methylhexan-2-one
:
:114.19 g/mol
EINECS:219-846-9
UN:1224
:GHS02
CAS Number:2550-21-2
Formula:C₇H₁₄O
MDL Number:MFCD00039948
Boiling Pt:145°C
Flash Pt:38°C
ADR:3 III