Order Entry
Italy
ContactUsLinkComponent
2-Methyl-3-heptanone ≥95.0% (by GC)
2-Methyl-3-heptanone ≥95.0% (by GC)
  TCIAM0693-5ML
 :  TCI
 :  
undefined
2-Methyl-3-heptanone ≥95.0% (by GC)
  TCIAM0693-5ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    5 mL
  • Seq:
    2210
  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 95.0 %

 

 

 : Butyl Isopropyl ketone, 2-Methylheptan-3-one
 :
 :128.21 g/mol
 :235-877-0
 :1224
 :GHS02
 :GHS07
 :13019-20-0
 :C₈H₁₆O
 :MFCD00009481
 :160°C
 :-59
 :43°C
 :3 III