n-Triacontane ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:638-68-6
:
Formula:H₃C(CH₂)₂₈CH₃
MDL Number:MFCD00009410
Boiling Pt:200°C
Molecular Weight:422.82 g/mol
Storage Temperature:Ambient
EINECS:211-349-5
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White to Almost white powder to crystal
- Purity (GC):min. 98.0 %
- Melting point:63.0 to 67.0 deg-C
- Solubility in hot Toluene:almost transparency
Specification Test Results
Frequently Bought Together