tert-Butyl acetate ≥99.0% (by GC)
: TCI
: 1,1-Dimethylethyl acetate
:540-88-5
:
:C₆H₁₂O₂
:MFCD00008807
:96°C
:116.16 g/mol
:15°C
:3 II
:14,01537
:208-760-7
:1123
:GHS02
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.0 %
- Acid Value:max. 1.0
Frequently Bought Together