Ethyl cinnamate ≥99.0% (by GC)
Supplier: TCI
Certificates
About this item
Synonyms: NSC 6773, 3-Phenyl-acrylicacidethylester, Ethyl 3-phenyl-2-propenoate, Cinnamic acid ethyl ester, 3-Phenyl-2-propenoic acid ethyl ester
CAS Number:103-36-6
Formula:C₁₁H₁₂O₂
MDL Number:MFCD00009189
Boiling Pt:271°C
Molecular Weight:176.21 g/mol
Melting Pt:10°C
Flash Pt:137°C
EINECS:203-104-6
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together