Order Entry
Italy
ContactUsLinkComponent
 
N-(3-(Trimethoxysilyl)propyl)ethylenediamine ≥97.0% (by GC)
 :  TCI
 
undefined
N-(3-(Trimethoxysilyl)propyl)ethylenediamine ≥97.0% (by GC)
 :  TCI

 

  • Environmentally Preferable:

 

 

  • Appearance:
    Colorless to Almost colorless clear liquid
  • Purity (GC):
    min. 97.0 %

 

Frequently Bought Together