Aluminium stearate, tech.
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Aluminum stearate, Aluminum tristearate
CAS Number:637-12-7
:
Formula:C₅₄H₁₀₅AlO₆
MDL Number:MFCD00036389
Molecular Weight:877.389 g/mol
Melting Pt:105…115 °C
Merck Index:14,364
Storage Temperature:Ambient
EINECS:211-279-5
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together