Nalidixic acid 99.5%
:389-08-2
:
:C₁₂H₁₂N₂O₃
:MFCD00006884
:232.24 g/mol
:225…231 °C
:206-864-7
:GHS08
:GHS07
:Ambient
- Environmentally Preferable:
- Appearance:White to light yellow Powder
- Infrared spectrum:Conforms
- Melting point:225 to 231 °C
- Titration with NaOH:≥99.4 %
- Specific optical rotation:-44.0° to -48.0° (20 °C, 589 nm) (c=0.7, water)
Frequently Bought Together