Proflavine hydrochloride (3,6-diaminoacridine hydrochloride) 95% (dye content), Sigma-Aldrich®
131105-25G
:
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
- Appearance:Dark orange to orange-brown powder
- Infrared spectrum:Conforms to structure.
- Uv-visible spectrum:In acidified methanol
- Carbon:59.7%-67.3%
- Nitrogen:15.9%-18.1%
- Titration:Report % Cl (with silver nitrate)
- High pressure liquid:94.0% (minimum)
- Solubility:1 mg/ml, H2O; clear, orange to brown
: Acridine-3,6-diamine hydrochloride
:
:245.71 g/mol
:270 °C (dec., lit.)
:Room Temperature
:213-459-9
:952-23-8
:C₁₃H₁₁N₃·HCl
:MFCD00066867